siRNA Details
| Gene Symbol: Huwe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021523.4 | NP_067498.4 | 39 | 5368 | ||
| GenBank mRNA | AB093227.2 | - | 5 | 583 | ||
| AY772010.3 | - | 39 | 5305 | |||
| AY929611.1 | - | 37 | 4915 | |||
| BC070444.1 | - | 1 | 29 | |||
| DQ097265.1 | - | 39 | 5324 | |||

Huwe1
Genomic Map