siRNA Details
| Gene Symbol: Mettl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021554.2 | NP_067529.2 | 2 | 637 | ||
| GenBank mRNA | AB041664.1 | - | 2 | 415 | ||
| AJ278576.1 | - | 2 | 637 | |||
| AK075586.1 | - | 2 | 400 | |||
| AK169951.1 | - | 1 | 53 | |||
| AK201397.1 | - | 2 | 125 | |||
| BC023188.1 | - | 1 | 78 | |||
| BC068124.1 | - | 2 | 431 | 5716972 | ||
| BC082317.1 | - | 1 | 62 | |||

Mettl9
Genomic Map