siRNA Details
| Gene Symbol: Bhlhe22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021560.4 | NP_067535.3 | 1 | 1473 | ||
| GenBank mRNA | AK047201.1 | - | 1 | 1207 | ||
| AK076228.1 | - | 1 | 1397 | |||
| BC053007.1 | - | 1 | 1473 | 5686844 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bhlhe22| Gene Symbol: Bhlhe22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021560.4 | NP_067535.3 | 1 | 1473 | ||
| GenBank mRNA | AK047201.1 | - | 1 | 1207 | ||
| AK076228.1 | - | 1 | 1397 | |||
| BC053007.1 | - | 1 | 1473 | 5686844 | ||