siRNA Details
| Gene Symbol: Ndufs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030274.1 | NP_001025445.1 | 2 | 271 | ||
| GenBank mRNA | AK148505.1 | - | 2 | 270 | ||
| AK152845.1 | - | 2 | 266 | |||
| BC081434.1 | - | 2 | 215 | 6412214 | ||
| BU534760.1 | - | 2 | 274 | |||
| Gene Symbol: BC002163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC002163.1 | - | 1 | 258 | ||

Ndufs5
Genomic Map