siRNA Details
| Gene Symbol: Il1rapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030688.1 | NP_109613.1 | 10 | 1393 | ||
| GenBank mRNA | AF284437.1 | - | 10 | 1393 | ||
| AK045976.1 | - | 2182 | ||||
| BC119274.1 | - | 10 | 1336 | 8734023 | ||
| BC120555.1 | - | 10 | 1336 | 40129478 | ||

Il1rapl2
Genomic Map