siRNA Details
| Gene Symbol: Il1rapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030688.1 | NP_109613.1 | 5, 6 | 807 | ||
| GenBank mRNA | AF284437.1 | - | 5, 6 | 807 | ||
| AJ277831.1 | - | 1565 | ||||
| AK039155.1 | - | 1332 | ||||
| AK045976.1 | - | 1596 | ||||
| BC119274.1 | - | 5, 6 | 750 | 8734023 | ||
| BC120555.1 | - | 5, 6 | 750 | 40129478 | ||

Il1rapl2
Genomic Map