siRNA Details
| Gene Symbol: Fam71a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109759.1 | NP_001103229.1 | 1 | 1478 | ||
| GenBank mRNA | AB473728.1 | - | 1 | 1105 | ||
| AK133412.1 | - | 1 | 1480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam71a| Gene Symbol: Fam71a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109759.1 | NP_001103229.1 | 1 | 1478 | ||
| GenBank mRNA | AB473728.1 | - | 1 | 1105 | ||
| AK133412.1 | - | 1 | 1480 | |||