siRNA Details
| Gene Symbol: Cldn34b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033799.2 | NP_001028971.1 | 3 | 648 | ||
| GenBank mRNA | AK132827.1 | - | 3 | 649 | ||
| BC147626.1 | - | 3 | 518 | 9056616 | ||
| BC147629.1 | - | 3 | 546 | 9056619 | ||
| BC172110.1 | - | 3 | 518 | 9054396 | ||

Cldn34b4
Genomic Map