siRNA Details
Gene Symbol: Zfp872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033813.4 | NP_001028985.1 | 4 | 330 | ||
GenBank mRNA | AK137156.1 | - | 4 | 332 | ||
BC150971.1 | - | 4 | 323 | 9087884 | ||
BC151175.1 | - | 4 | 323 | 9088064 |
