siRNA Details
| Gene Symbol: Zcchc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153160.4 | NP_694800.1 | 6 | 461 | ||
| GenBank mRNA | AJ272345.1 | - | 6 | 417 | ||
| AK152289.1 | - | 6 | 462 | |||
| AK161588.1 | - | 6 | 456 | |||
| AY253297.1 | - | 6 | 470 | |||
| BC018382.1 | - | 5 | 399 | 4215774 | ||
| BC049231.1 | - | 6 | 437 | 4161178 | ||

Zcchc17
Genomic Map