siRNA Details
| Gene Symbol: Klf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135093.1 | NP_001128565.1 | 1 | 999 | ||
| GenBank mRNA | AK030435.1 | - | 2 | 293 | ||
| DQ534758.1 | - | 1 | 991 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf14| Gene Symbol: Klf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135093.1 | NP_001128565.1 | 1 | 999 | ||
| GenBank mRNA | AK030435.1 | - | 2 | 293 | ||
| DQ534758.1 | - | 1 | 991 | |||