siRNA Details
| Gene Symbol: Catsperd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175350.3 | NP_780559.2 | 7 | 546 | ||
| GenBank mRNA | HQ441159.1 | - | 7 | 538 | ||
| HQ441160.1 | - | 5 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Catsperd| Gene Symbol: Catsperd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175350.3 | NP_780559.2 | 7 | 546 | ||
| GenBank mRNA | HQ441159.1 | - | 7 | 538 | ||
| HQ441160.1 | - | 5 | 320 | |||