siRNA Details
| Gene Symbol: Gpr52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146330.1 | NP_001139802.1 | 1 | 271 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr52| Gene Symbol: Gpr52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146330.1 | NP_001139802.1 | 1 | 271 | ||
Set Membership: |
|