siRNA Details
| Gene Symbol: Fam150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195732.1 | NP_001182661.1 | 2 | 204 | ||
| GenBank mRNA | AK087278.1 | - | 2 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam150a| Gene Symbol: Fam150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195732.1 | NP_001182661.1 | 2 | 204 | ||
| GenBank mRNA | AK087278.1 | - | 2 | 80 | ||