siRNA Details
| Gene Symbol: Zfp963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200023.1 | NP_001186952.1 | 4 | 1055 | ||
| GenBank mRNA | AK162670.1 | - | 4 | 1056 | ||
| BC037669.1 | - | 2 | 767 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp963| Gene Symbol: Zfp963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200023.1 | NP_001186952.1 | 4 | 1055 | ||
| GenBank mRNA | AK162670.1 | - | 4 | 1056 | ||
| BC037669.1 | - | 2 | 767 | |||