siRNA Details
| Gene Symbol: Gm12185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045540.2 | NP_001039005.1 | 2 | 874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12185| Gene Symbol: Gm12185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045540.2 | NP_001039005.1 | 2 | 874 | ||