siRNA Details
| Gene Symbol: Nhlrc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039038.2 | NP_001034127.1 | 2 | 887 | ||
| GenBank mRNA | AK143822.1 | - | 2 | 887 | ||
| BC147284.1 | - | 1 | 477 | 9056317 | ||
| BC147285.1 | - | 1 | 476 | 9056337 | ||

Nhlrc4
Genomic Map