siRNA Details
| Gene Symbol: Gm10244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB106361.1 | - | 1, 2 | 272 | ||
| AB106871.1 | - | 1, 2 | 272 | |||
| AK076873.1 | - | 1, 2 | 272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10244| Gene Symbol: Gm10244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB106361.1 | - | 1, 2 | 272 | ||
| AB106871.1 | - | 1, 2 | 272 | |||
| AK076873.1 | - | 1, 2 | 272 | |||