siRNA Details
Gene Symbol: Gm9785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147682.1 | - | 1 | 426 | 9056655 | |
BC147695.1 | - | 1 | 426 | 9056668 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147682.1 | - | 1 | 426 | 9056655 | |
BC147695.1 | - | 1 | 426 | 9056668 |