siRNA Details
| Gene Symbol: Kctd21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039039.3 | NP_001034128.2 | 2 | 282 | ||
| GenBank mRNA | AK141547.1 | - | 2 | 279 | ||
| BC139363.1 | - | 1 | 231 | 8862385 | ||
| BC139365.1 | - | 1 | 231 | 8862387 | ||

Kctd21
Genomic Map