siRNA Details
| Gene Symbol: Arhgef26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081295.1 | NP_001074764.1 | 6 | 2254 | ||
| GenBank mRNA | AK144976.1 | - | 6 | 1760 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef26| Gene Symbol: Arhgef26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081295.1 | NP_001074764.1 | 6 | 2254 | ||
| GenBank mRNA | AK144976.1 | - | 6 | 1760 | ||