siRNA Details
| Gene Symbol: 1700123I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165919.1 | NP_001159391.1 | 5 | 437 | ||
| GenBank mRNA | BC138005.1 | - | 5 | 351 | 8861024 | |
| BC145092.1 | - | 5 | 351 | 9053640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700123I01Rik| Gene Symbol: 1700123I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165919.1 | NP_001159391.1 | 5 | 437 | ||
| GenBank mRNA | BC138005.1 | - | 5 | 351 | 8861024 | |
| BC145092.1 | - | 5 | 351 | 9053640 | ||