siRNA Details
Gene Symbol: Tmem200c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206661.1 | NP_001193590.1 | 2 | 1481 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem200c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206661.1 | NP_001193590.1 | 2 | 1481 |