siRNA Details
Gene Symbol: Zfp827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294279.1 | NP_001281208.1 | 2 | 1160 | ||
NM_178267.3 | NP_839998.2 | 2 | 1160 | |||
GenBank mRNA | BC094552.1 | - | 2 | 1160 | ||
BC151183.1 | - | 2 | 1045 | 9088072 | ||
CF539233.1 | - | 1 | 342 |
