siRNA Details
| Gene Symbol: Nme9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165957.1 | NP_001159429.1 | 6 | 377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nme9| Gene Symbol: Nme9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165957.1 | NP_001159429.1 | 6 | 377 | ||