siRNA Details
| Gene Symbol: Lipt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037918.3 | NP_001033007.2 | 2 | 677 | ||
| GenBank mRNA | AK048005.1 | - | 678 | |||
| BC019519.1 | - | 2 | 666 | 4038179 | ||
| DV649848.1 | - | 2 | 609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lipt1| Gene Symbol: Lipt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037918.3 | NP_001033007.2 | 2 | 677 | ||
| GenBank mRNA | AK048005.1 | - | 678 | |||
| BC019519.1 | - | 2 | 666 | 4038179 | ||
| DV649848.1 | - | 2 | 609 | |||