siRNA Details
| Gene Symbol: Btnl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030747.1 | NP_109672.1 | 3 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btnl6| Gene Symbol: Btnl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030747.1 | NP_109672.1 | 3 | 653 | ||