siRNA Details
| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037717.3 | NP_001032806.2 | 7 | 592 | ||
| GenBank mRNA | BC157917.1 | - | 7 | 541 | 9007345 | |
| BC157928.1 | - | 7 | 541 | 9088101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc38a6| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037717.3 | NP_001032806.2 | 7 | 592 | ||
| GenBank mRNA | BC157917.1 | - | 7 | 541 | 9007345 | |
| BC157928.1 | - | 7 | 541 | 9088101 | ||