siRNA Details
| Gene Symbol: Tmem236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081310.2 | NP_001074779.1 | 4 | 1072 | ||
| GenBank mRNA | AK144489.1 | - | 4 | 1074 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem236| Gene Symbol: Tmem236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081310.2 | NP_001074779.1 | 4 | 1072 | ||
| GenBank mRNA | AK144489.1 | - | 4 | 1074 | ||