siRNA Details
| Gene Symbol: Gm6588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177504.1 | NP_001170975.1 | 1 | 545 | ||
| GenBank mRNA | AK133454.1 | - | 1 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6588| Gene Symbol: Gm6588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177504.1 | NP_001170975.1 | 1 | 545 | ||
| GenBank mRNA | AK133454.1 | - | 1 | 545 | ||