siRNA Details
| Gene Symbol: AB041806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB041806.1 | - | 1 | 330 | ||
| Gene Symbol: Fam43b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081672.2 | NP_001075141.1 | 1 | 928 | ||

AB041806
Genomic Map