siRNA Details
| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087784.1 | - | 2 | 207 | ||
| AK143622.1 | - | 2 | 209 | |||
| AK154515.1 | - | 431 | ||||
| AK168480.1 | - | 2 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E330020D12Rik| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087784.1 | - | 2 | 207 | ||
| AK143622.1 | - | 2 | 209 | |||
| AK154515.1 | - | 431 | ||||
| AK168480.1 | - | 2 | 207 | |||