siRNA Details
Gene Symbol: Vmn2r93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104542.1 | NP_001098012.1 | 3 | 1258 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
