siRNA Details
| Gene Symbol: Vmn2r93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104542.1 | NP_001098012.1 | 6 | 1857 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r93| Gene Symbol: Vmn2r93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104542.1 | NP_001098012.1 | 6 | 1857 | ||
Set Membership: |
|