siRNA Details
Gene Symbol: Fam196a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143802.1 | NP_001137274.1 | 3 | 1938 | ||
GenBank mRNA | AK046846.1 | - | 2 | 654 | ||
AK140373.1 | - | 3 | 1937 | |||
BC158077.1 | - | 4 | 1561 | 9088169 | ||
BC158080.1 | - | 4 | 1561 | 9088172 |
