siRNA Details
| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195075.1 | NP_001182004.1 | 10 | 929 | ||
| GenBank mRNA | CK780311.1 | - | 4 | 343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl3| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195075.1 | NP_001182004.1 | 10 | 929 | ||
| GenBank mRNA | CK780311.1 | - | 4 | 343 | ||