siRNA Details
| Gene Symbol: Gm6792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177416.1 | NP_001170887.1 | 1 | 110 | ||
| GenBank mRNA | AY226991.1 | - | 1 | 109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6792| Gene Symbol: Gm6792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177416.1 | NP_001170887.1 | 1 | 110 | ||
| GenBank mRNA | AY226991.1 | - | 1 | 109 | ||