siRNA Details
| Gene Symbol: Gm6792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177416.1 | NP_001170887.1 | 10, 12 | 347 | ||
| NM_001177417.1 | NP_001170888.1 | 9, 11 | 166 | |||
| GenBank mRNA | AK079003.1 | - | 8, 10 | 171 | ||
| AY226991.1 | - | 10, 12 | 345 | |||

Gm6792
Genomic Map