siRNA Details
| Gene Symbol: Gm11437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037932.2 | NP_001033021.2 | 5, 6 | 773 | ||
| GenBank mRNA | AJ844653.1 | - | 5, 6 | 773 | ||
| BC147775.1 | - | 5, 6 | 765 | 9056710 | ||
| BC147791.1 | - | 5, 6 | 765 | 9056714 | ||

Gm11437
Genomic Map