siRNA Details
| Gene Symbol: Dact3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081655.1 | NP_001075124.1 | 4 | 656 | ||
| GenBank mRNA | AB073971.1 | - | 4 | 577 | ||
| DQ832319.1 | - | 4 | 656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dact3| Gene Symbol: Dact3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081655.1 | NP_001075124.1 | 4 | 656 | ||
| GenBank mRNA | AB073971.1 | - | 4 | 577 | ||
| DQ832319.1 | - | 4 | 656 | |||