siRNA Details
| Gene Symbol: Mroh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039557.4 | NP_001034646.2 | 19 | 2708 | ||
| GenBank mRNA | AK029507.1 | - | 19 | 2680 | ||
| AK132763.1 | - | 19 | 2465 | |||
| BC150937.1 | - | 19 | 2432 | 9087848 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mroh8| Gene Symbol: Mroh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039557.4 | NP_001034646.2 | 19 | 2708 | ||
| GenBank mRNA | AK029507.1 | - | 19 | 2680 | ||
| AK132763.1 | - | 19 | 2465 | |||
| BC150937.1 | - | 19 | 2432 | 9087848 | ||