siRNA Details
| Gene Symbol: Wfdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039501.2 | NP_001034590.1 | 1, 2 | 149 | ||
| GenBank mRNA | AK131754.1 | - | 1, 2 | 150 | ||
| BC132270.1 | - | 1, 2 | 140 | 40130547 | ||
| BC132272.1 | - | 1, 2 | 140 | 40130549 | ||
| DQ437333.1 | - | 1, 2 | 255 | |||

Wfdc10
Genomic Map