siRNA Details
| Gene Symbol: Wfdc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161806.1 | NP_001155278.1 | 4 | 376 | ||
| GenBank mRNA | AK079019.1 | - | 4 | 377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc11| Gene Symbol: Wfdc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161806.1 | NP_001155278.1 | 4 | 376 | ||
| GenBank mRNA | AK079019.1 | - | 4 | 377 | ||