siRNA Details
| Gene Symbol: Gm7008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004585.1 | - | 715 | |||
| AK016351.1 | - | 811 | ||||
| CB273562.1 | - | 2 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7008| Gene Symbol: Gm7008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004585.1 | - | 715 | |||
| AK016351.1 | - | 811 | ||||
| CB273562.1 | - | 2 | 207 | |||