siRNA Details
Gene Symbol: Cyp4f40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101588.1 | NP_001095058.1 | 1 | 115 | ||
GenBank mRNA | BC158089.1 | - | 1 | 192 | 9088175 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp4f40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101588.1 | NP_001095058.1 | 1 | 115 | ||
GenBank mRNA | BC158089.1 | - | 1 | 192 | 9088175 |