siRNA Details
| Gene Symbol: Ano8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042391.1 | - | 6 | 1439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano8| Gene Symbol: Ano8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042391.1 | - | 6 | 1439 | ||