siRNA Details
| Gene Symbol: Gm12250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135115.1 | NP_001128587.1 | 2 | 1061 | ||
| GenBank mRNA | DQ508486.1 | - | 1 | 799 | ||
| DQ508487.1 | - | 1 | 799 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12250| Gene Symbol: Gm12250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135115.1 | NP_001128587.1 | 2 | 1061 | ||
| GenBank mRNA | DQ508486.1 | - | 1 | 799 | ||
| DQ508487.1 | - | 1 | 799 | |||