siRNA Details
Gene Symbol: Dmrtc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039116.2 | NP_001034205.2 | 2 | 183 | ||
GenBank mRNA | BC150829.1 | - | 2 | 183 | 9087740 | |
DQ316775.1 | - | 1 | 160 | |||
DQ316776.1 | - | 1 | 320 | |||
DQ316777.1 | - | 2 | 238 |
