siRNA Details
| Gene Symbol: Gm7120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039244.3 | NP_001034333.2 | 2, 3 | 542 | ||
| NM_001177666.1 | NP_001171137.1 | 2, 3 | 515 | |||
| GenBank mRNA | AK050607.1 | - | 2, 3 | 543 | ||
| BC021422.1 | - | 2, 3 | 524 | 5011629 | ||
| BC103784.1 | - | 1, 2 | 518 | 6309338 | ||

Gm7120
Genomic Map