siRNA Details
| Gene Symbol: Gm7173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099307.1 | NP_001092777.1 | 2 | 261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7173| Gene Symbol: Gm7173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099307.1 | NP_001092777.1 | 2 | 261 | ||